Anti-KYNU Mouse Monoclonal Antibody [clone: 589731]

Supplier: R & D Systems
MAB4887
102885-828EA 577.02 USD
102885-828
Anti-KYNU Mouse Monoclonal Antibody [clone: 589731]
Antibodies
Type: Primary
Antigen: KYNU
Clonality: Monoclonal
Clone: 589731
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR