Anti-THPO Rat Monoclonal Antibody [clone: 378120]

Supplier: R & D Systems
MAB488
220021-047EA 714.4 USD
220021-047
Anti-THPO Rat Monoclonal Antibody [clone: 378120]
Antibodies
Type: Primary
Antigen: THPO
Clonality: Monoclonal
Clone: 378120
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR