Anti-SCGN Mouse Monoclonal Antibody [clone: 778518]

Supplier: R & D Systems
MAB4878
102885-826EA 594.37 USD
102885-826
Anti-SCGN Mouse Monoclonal Antibody [clone: 778518]
Antibodies
Simple Western

Type: Primary
Antigen: SCGN
Clonality: Monoclonal
Clone: 778518
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR