Anti-S100 A11 Mouse Monoclonal Antibody [clone: 672816]

Supplier: R & D Systems
MAB4874
102885-824EA 577.02 USD
102885-824
Anti-S100 A11 Mouse Monoclonal Antibody [clone: 672816]
Antibodies
Type: Primary
Antigen: S100 A11
Clonality: Monoclonal
Clone: 672816
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR