Anti-CCL6 Rat Monoclonal Antibody [clone: 262015]

Supplier: R & D Systems
MAB4871
102885-822EA 763.57 USD
102885-822
Anti-CCL6 Rat Monoclonal Antibody [clone: 262015]
Antibodies
Type: Primary
Antigen: CCL6
Clonality: Monoclonal
Clone: 262015
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR