Anti-S100 A2 Mouse Monoclonal Antibody [clone: 674033]

Supplier: R & D Systems
MAB48701
102885-820EA 577.02 USD
102885-820
Anti-S100 A2 Mouse Monoclonal Antibody [clone: 674033]
Antibodies
Type: Primary
Antigen: S100 A2
Clonality: Monoclonal
Clone: 674033
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR