Anti-CCL6 Rat Monoclonal Antibody [clone: 262016]

Supplier: R & D Systems
MAB487
220021-046EA 779.48 USD
220021-046
Anti-CCL6 Rat Monoclonal Antibody [clone: 262016]
Antibodies
Type: Primary
Antigen: CCL6
Clonality: Monoclonal
Clone: 262016
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR