Anti-HLA-DRA Mouse Monoclonal Antibody [clone: L203]

Supplier: R & D Systems
MAB4869
220021-045EA 594.37 USD
220021-045
Anti-HLA-DRA Mouse Monoclonal Antibody [clone: L203]
Antibodies
Type: Primary
Antigen: HLA-DRA
Clonality: Monoclonal
Clone: L203
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR