Anti-AATK Mouse Monoclonal Antibody [clone: 4566]

Supplier: R & D Systems
MAB4866
220021-042EA 561.11 USD
220021-042
Anti-AATK Mouse Monoclonal Antibody [clone: 4566]
Antibodies
Type: Primary
Antigen: AATK
Clonality: Monoclonal
Clone: 4566
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR