Anti-CD81 Rat Monoclonal Antibody [clone: 431301]

Supplier: R & D Systems
MAB4865
220021-041EA 594.37 USD
220021-041
Anti-CD81 Rat Monoclonal Antibody [clone: 431301]
Antibodies
Type: Primary
Antigen: CD81
Clonality: Monoclonal
Clone: 431301
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR