Anti-CYR61 Rat Monoclonal Antibody [clone: 466305]

Supplier: R & D Systems
MAB4864
220021-040EA 577.02 USD
220021-040
Anti-CYR61 Rat Monoclonal Antibody [clone: 466305]
Antibodies
Type: Primary
Antigen: CYR61
Clonality: Monoclonal
Clone: 466305
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR