Anti-NPR1 Mouse Monoclonal Antibody [clone: 377029]

Supplier: R & D Systems
MAB4860
220021-039EA 594.37 USD
220021-039
Anti-NPR1 Mouse Monoclonal Antibody [clone: 377029]
Antibodies
Type: Primary
Antigen: NPR1
Clonality: Monoclonal
Clone: 377029
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR