Anti-XCL1 Rat Monoclonal Antibody [clone: 80222]

Supplier: R & D Systems
MAB486
220021-038EA 779.48 USD
220021-038
Anti-XCL1 Rat Monoclonal Antibody [clone: 80222]
Antibodies
Type: Primary
Antigen: XCL1
Clonality: Monoclonal
Clone: 80222
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR