Anti-PIK3AP1 Mouse Monoclonal Antibody [clone: 501813]

Supplier: R & D Systems
MAB4857
220021-037EA 594.37 USD
220021-037
Anti-PIK3AP1 Mouse Monoclonal Antibody [clone: 501813]
Antibodies
Type: Primary
Antigen: PIK3AP1
Clonality: Monoclonal
Clone: 501813
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR