Anti-ANXA3 Mouse Monoclonal Antibody [clone: 69351]

Supplier: R & D Systems
MAB4855
102885-816EA 577.02 USD
102885-816
Anti-ANXA3 Mouse Monoclonal Antibody [clone: 69351]
Antibodies
Type: Primary
Antigen: ANXA3
Clonality: Monoclonal
Clone: 69351
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR