Anti-MALT1 Mouse Monoclonal Antibody [clone: MALT1-C]

Supplier: R & D Systems
MAB4850
220021-035EA 623.29 USD
220021-035
Anti-MALT1 Mouse Monoclonal Antibody [clone: MALT1-C]
Antibodies
Simple Western

Type: Primary
Antigen: MALT1
Clonality: Monoclonal
Clone: MALT1-C
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR