Anti-CRYAB Mouse Monoclonal Antibody [clone: 731502]

Supplier: R & D Systems
MAB4849
102885-814EA 594.37 USD
102885-814
Anti-CRYAB Mouse Monoclonal Antibody [clone: 731502]
Antibodies
Simple Western

Type: Primary
Antigen: CRYAB
Clonality: Monoclonal
Clone: 731502
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR