Anti-ISG15 Mouse Monoclonal Antibody [clone: 539442]

Supplier: R & D Systems
MAB4845
220021-032EA 594.37 USD
220021-032
Anti-ISG15 Mouse Monoclonal Antibody [clone: 539442]
Antibodies
Type: Primary
Antigen: ISG15
Clonality: Monoclonal
Clone: 539442
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR