Anti-INA Mouse Monoclonal Antibody [clone: 529724]

Supplier: R & D Systems
MAB4844
220021-031EA 561.11 USD
220021-031
Anti-INA Mouse Monoclonal Antibody [clone: 529724]
Antibodies
Type: Primary
Antigen: INA
Clonality: Monoclonal
Clone: 529724
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR