Anti-GRM1 Mouse Monoclonal Antibody [clone: 511622]

Supplier: R & D Systems
MAB4836
220021-028EA 561.11 USD
220021-028
Anti-GRM1 Mouse Monoclonal Antibody [clone: 511622]
Antibodies
Type: Primary
Antigen Symbol: GRM1
Antigen Name: glutamate receptor, metabotropic 1
Antigen Synonyms: GPRC1A, MGLU1, MGLUR1, PPP1R85, SCAR13
Clonality: Monoclonal
Clone: 511622
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR