Anti-CD28 Rat Monoclonal Antibody [clone: 794716]

Supplier: R & D Systems
MAB4832
102885-812EA 655.11 USD
102885-812
Anti-CD28 Rat Monoclonal Antibody [clone: 794716]
Antibodies
Type: Primary
Antigen: CD28
Clonality: Monoclonal
Clone: 794716
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR