Anti-IRF1 Mouse Monoclonal Antibody [clone: 686703]

Supplier: R & D Systems
MAB4830
102885-810EA 577.02 USD
102885-810
Anti-IRF1 Mouse Monoclonal Antibody [clone: 686703]
Antibodies
Type: Primary
Antigen: IRF1
Clonality: Monoclonal
Clone: 686703
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR