Anti-CTBS Mouse Monoclonal Antibody [clone: 486725]

Supplier: R & D Systems
MAB4829
220021-026EA 561.11 USD
220021-026
Anti-CTBS Mouse Monoclonal Antibody [clone: 486725]
Antibodies
Type: Primary
Antigen: CTBS
Clonality: Monoclonal
Clone: 486725
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR