Anti-RET Rat Monoclonal Antibody [clone: 64623]

Supplier: R & D Systems
MAB482
220021-025EA 772.25 USD
220021-025
Anti-RET Rat Monoclonal Antibody [clone: 64623]
Antibodies
Type: Primary
Antigen: RET
Clonality: Monoclonal
Clone: 64623
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR