Anti-VSIG1 Mouse Monoclonal Antibody [clone: 771506]

Supplier: R & D Systems
MAB4818
102885-808EA 616.06 USD
102885-808
Anti-VSIG1 Mouse Monoclonal Antibody [clone: 771506]
Antibodies
Simple Western

Type: Primary
Antigen: VSIG1
Clonality: Monoclonal
Clone: 771506
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR