Anti-CCL25 Rat Monoclonal Antibody [clone: 89827]

Supplier: R & D Systems
MAB4811
220021-024EA 763.57 USD
220021-024
Anti-CCL25 Rat Monoclonal Antibody [clone: 89827]
Antibodies
Type: Primary
Antigen: CCL25
Clonality: Monoclonal
Clone: 89827
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR