Anti-IL1RN Rat Monoclonal Antibody [clone: 694204]

Supplier: R & D Systems
MAB4801
102885-806EA 577.02 USD
102885-806
Anti-IL1RN Rat Monoclonal Antibody [clone: 694204]
Antibodies
Type: Primary
Antigen: IL1RN
Clonality: Monoclonal
Clone: 694204
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR