Anti-LAMP1 Mouse Monoclonal Antibody [clone: 508921]

Supplier: R & D Systems
MAB4800
102885-804EA 639.2 USD
102885-804
Anti-LAMP1 Mouse Monoclonal Antibody [clone: 508921]
Antibodies
Type: Primary
Antigen: LAMP1
Clonality: Monoclonal
Clone: 508921
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR