Anti-IL1RN Rat Monoclonal Antibody [clone: 40007]

Supplier: R & D Systems
MAB480
220021-022EA 772.25 USD
220021-022
Anti-IL1RN Rat Monoclonal Antibody [clone: 40007]
Antibodies
Type: Primary
Antigen: IL1RN
Clonality: Monoclonal
Clone: 40007
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR