Anti-DOK3 Mouse Monoclonal Antibody [clone: 642316]

Supplier: R & D Systems
MAB4795
102885-800EA 561.11 USD
102885-800
Anti-DOK3 Mouse Monoclonal Antibody [clone: 642316]
Antibodies
Type: Primary
Antigen: DOK3
Clonality: Monoclonal
Clone: 642316
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR