Anti-IGSF3 Mouse Monoclonal Antibody [clone: 503621]

Supplier: R & D Systems
MAB4788
220021-020EA 577.02 USD
220021-020
Anti-IGSF3 Mouse Monoclonal Antibody [clone: 503621]
Antibodies
Type: Primary
Antigen: IGSF3
Clonality: Monoclonal
Clone: 503621
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR