Anti-ADRBK2 Mouse Monoclonal Antibody [clone: 49718]

Supplier: R & D Systems
MAB4785
220021-019EA 561.11 USD
220021-019
Anti-ADRBK2 Mouse Monoclonal Antibody [clone: 49718]
Antibodies
Type: Primary
Antigen: ADRBK2
Clonality: Monoclonal
Clone: 49718
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR