Anti-CFH Mouse Monoclonal Antibody [clone: 556317]

Supplier: R & D Systems
MAB4779
102885-798EA 600.15 USD
102885-798
Anti-CFH Mouse Monoclonal Antibody [clone: 556317]
Antibodies
Type: Primary
Antigen: CFH
Clonality: Monoclonal
Clone: 556317
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR