Anti-HPN Mouse Monoclonal Antibody [clone: 506230]

Supplier: R & D Systems
MAB47761
220021-016EA 594.37 USD
220021-016
Anti-HPN Mouse Monoclonal Antibody [clone: 506230]
Antibodies
Type: Primary
Antigen: HPN
Clonality: Monoclonal
Clone: 506230
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR