Anti-HPN Mouse Monoclonal Antibody [clone: 506220]

Supplier: R & D Systems
MAB4776
220021-015EA 594.37 USD
220021-015
Anti-HPN Mouse Monoclonal Antibody [clone: 506220]
Antibodies
Type: Primary
Antigen: HPN
Clonality: Monoclonal
Clone: 506220
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR