Anti-IL7R Rat Monoclonal Antibody [clone: 132206]

Supplier: R & D Systems
MAB4774
220021-013EA 763.57 USD
220021-013
Anti-IL7R Rat Monoclonal Antibody [clone: 132206]
Antibodies
Type: Primary
Antigen: IL7R
Clonality: Monoclonal
Clone: 132206
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR