Anti-PODXL Mouse Monoclonal Antibody [clone: TRA-1-60]

Supplier: R & D Systems
MAB4770
220021-012EA 647.88 USD
220021-012
Anti-PODXL Mouse Monoclonal Antibody [clone: TRA-1-60]
Antibodies
Type: Primary
Antigen: PODXL
Clonality: Monoclonal
Clone: TRA-1-60
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR