Anti-VSIG2 Rat Monoclonal Antibody [clone: 771625]

Supplier: R & D Systems
MAB4766
102885-796EA 566.89 USD
102885-796
Anti-VSIG2 Rat Monoclonal Antibody [clone: 771625]
Antibodies
Type: Primary
Antigen: VSIG2
Clonality: Monoclonal
Clone: 771625
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR