Anti-AZGP1 Mouse Monoclonal Antibody [clone: 842025]

Supplier: R & D Systems
MAB4764
102885-794EA 594.37 USD
102885-794
Anti-AZGP1 Mouse Monoclonal Antibody [clone: 842025]
Antibodies
Simple Western

Type: Primary
Antigen: AZGP1
Clonality: Monoclonal
Clone: 842025
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR