Anti-SCARA5 Rat Monoclonal Antibody [clone: 462002]

Supplier: R & D Systems
MAB4754
220021-010EA 594.37 USD
220021-010
Anti-SCARA5 Rat Monoclonal Antibody [clone: 462002]
Antibodies
Type: Primary
Antigen: SCARA5
Clonality: Monoclonal
Clone: 462002
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR