Anti-WNT4 Rat Monoclonal Antibody [clone: 55025]

Supplier: R & D Systems
MAB4751
220021-009EA 594.37 USD
220021-009
Anti-WNT4 Rat Monoclonal Antibody [clone: 55025]
Antibodies
Type: Primary
Antigen: WNT4
Clonality: Monoclonal
Clone: 55025
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR