Anti-WNT4 Rat Monoclonal Antibody [clone: 55010]

Supplier: R & D Systems
MAB475-500 MAB475-050
220021-011EA 815.63 USD
220021-011 220021-008
Anti-WNT4 Rat Monoclonal Antibody [clone: 55010]
Antibodies
Type: Primary
Antigen: WNT4
Clonality: Monoclonal
Clone: 55010
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR