Anti-JAG2 Rat Monoclonal Antibody [clone: 746513]

Supplier: R & D Systems
MAB4748
102885-792EA 566.89 USD
102885-792
Anti-JAG2 Rat Monoclonal Antibody [clone: 746513]
Antibodies
Type: Primary
Antigen: JAG2
Clonality: Monoclonal
Clone: 746513
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR