Anti-IL10RA Hamster Monoclonal Antibody [clone: 105653]

Supplier: R & D Systems
MAB474
220021-007EA 763.57 USD
220021-007
Anti-IL10RA Hamster Monoclonal Antibody [clone: 105653]
Antibodies
Type: Primary
Antigen: IL10RA
Clonality: Monoclonal
Clone: 105653
Conjugation: Unconjugated
Epitope:
Host: Hamster
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR