Anti-TSPAN8 Rat Monoclonal Antibody [clone: 458811]

Supplier: R & D Systems
MAB4734
220021-006EA 600.15 USD
220021-006
Anti-TSPAN8 Rat Monoclonal Antibody [clone: 458811]
Antibodies
Type: Primary
Antigen: TSPAN8
Clonality: Monoclonal
Clone: 458811
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR