Anti-EFNB1 Rat Monoclonal Antibody [clone: 94038]

Supplier: R & D Systems
MAB4731
220021-005EA 763.57 USD
220021-005
Anti-EFNB1 Rat Monoclonal Antibody [clone: 94038]
Antibodies
Type: Primary
Antigen: EFNB1
Clonality: Monoclonal
Clone: 94038
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR