Anti-C5AR2 Rat Monoclonal Antibody [clone: 468705]

Supplier: R & D Systems
MAB4729
220021-003EA 594.37 USD
220021-003
Anti-C5AR2 Rat Monoclonal Antibody [clone: 468705]
Antibodies
Type: Primary
Antigen: C5AR2
Clonality: Monoclonal
Clone: 468705
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR