Anti-NMBR Mouse Monoclonal Antibody [clone: 466501]

Supplier: R & D Systems
MAB47281
102885-790EA 566.89 USD
102885-790
Anti-NMBR Mouse Monoclonal Antibody [clone: 466501]
Antibodies
Type: Primary
Antigen: NMBR
Clonality: Monoclonal
Clone: 466501
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR