Anti-NMBR Mouse Monoclonal Antibody [clone: 466505]

Supplier: R & D Systems
MAB4728
220021-002EA 577.02 USD
220021-002
Anti-NMBR Mouse Monoclonal Antibody [clone: 466505]
Antibodies
Type: Primary
Antigen: NMBR
Clonality: Monoclonal
Clone: 466505
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR