Anti-MRGPRX2 Mouse Monoclonal Antibody [clone: 477533]

Supplier: R & D Systems
MAB4727
220021-001EA 616.06 USD
220021-001
Anti-MRGPRX2 Mouse Monoclonal Antibody [clone: 477533]
Antibodies
Type: Primary
Antigen Symbol: MRGPRX2
Antigen Name: MAS-related GPR, member X2
Antigen Synonyms: MGRG3, MRGX2
Clonality: Monoclonal
Clone: 477533
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR